3-(Trifluormethyl)anthranilsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Amino-α,α,α-trifluoro-m-toluic acid, 2-Amino-3-(trifluoromethyl)benzoic acid

AS33770G1 AS33770G10 AS33770G5
ACTIAS33770G1EA 0 CHF
ACTIAS33770G1 ACTIAS33770G10 ACTIAS33770G5
3-(Trifluormethyl)anthranilsäure
3-(Trifluormethyl)anthranilsäure
Formel: C₈H₆F₃NO₂
MDL Number: MFCD00274210
CAS-Nummer: 313-12-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR