α,α,α,4-Tetrafluor-m-toluylsäure

Lieferant: ACTIVATE SCIENTIFIC

AS35008G1 AS35008G10 AS35008G5
ACTIAS35008G1EA 0 CHF
ACTIAS35008G1 ACTIAS35008G10 ACTIAS35008G5
α,α,α,4-Tetrafluor-m-toluylsäure
α,α,α,4-Tetrafluor-m-toluylsäure
Formel: C₈H₄F₄O₂
MDL Number: MFCD00061294
CAS-Nummer: 67515-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR