3-Fluor-o-toluylsäure

Lieferant: ACTIVATE SCIENTIFIC

AS31343G1 AS31343G5
ACTIAS31343G1EA 0 CHF
ACTIAS31343G1 ACTIAS31343G5
3-Fluor-o-toluylsäure
3-Fluor-o-toluylsäure
Formel: C₈H₇FO₂
MW: 154,14 g/mol
MDL Number: MFCD00075246
CAS-Nummer: 699-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR