(2,4,5-Trifluorphenyl)acetonitril
Lieferant: ACTIVATE SCIENTIFIC
AS35145G1
AS35145G5
ACTIAS35145G1EA
0
CHF
ACTIAS35145G1
ACTIAS35145G5
(2,4,5-Trifluorphenyl)acetonitril
(2,4,5-Trifluorphenyl)acetonitril
Formel:
C₈H₄F₃N MW: 171,12 g/mol |
MDL Number:
MFCD00061220 CAS-Nummer: 220141-74-2 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...