(3-Chlorphenyl)acetonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Chlorobenzyl cyanide

AS21757G100 AS21757G25
ACTIAS21757G100EA 206 CHF
ACTIAS21757G100 ACTIAS21757G25
(3-Chlorphenyl)acetonitril
(3-Chlorphenyl)acetonitril
Formel: C₈H₆ClN
MW: 151.6 g/mol
Siedepunkt: 270
Melting Pt: 11,5
Dichte: 1.1806 g/cm³ (30 °C)
MDL Number: MFCD00001908
CAS-Nummer: 1529-41-5
EINECS: 216-213-9
UN: 3276
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR