ω-Brom-3-fluoracetophenon
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
Bromomethyl m-fluorophenyl ketone, 3-Fluorophenacyl bromide, 2-Bromo-1-(3-fluorophenyl)ethan-1-one, α-Bromo-3-fluoroacetophenone, 2-Bromo-3'-fluoroacetophenone
AS32795G1
AS32795G5
ACTIAS32795G1EA
0
CHF
ACTIAS32795G1
ACTIAS32795G5
ω-Brom-3-fluoracetophenon
ω-Brom-3-fluoracetophenon
Formel:
C₈H₆BrFO MW: 217,04 g/mol Lager Temperatur: Kühlschrank |
MDL Number:
MFCD00109756 CAS-Nummer: 53631-18-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...