(4-Bromphenyl)acetonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Bromophenylacetonitrile, 4-Bromobenzyl cyanide

AS33461G10 AS33461G100
ACTIAS33461G10EA 0 CHF
ACTIAS33461G10 ACTIAS33461G100
(4-Bromphenyl)acetonitril
(4-Bromphenyl)acetonitril
Formel: C₈H₆BrN
MW: 196.05 g/mol
Siedepunkt: 286
Melting Pt: 47…49 °C
MDL Number: MFCD00001916
CAS-Nummer: 16532-79-9
EINECS: 240-602-2
UN: 3449
ADR: 6.1 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR