1-Brom-4-propylbenzol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (4-Bromophenyl)propane, 1-Bromo-4-n-propylbenzene

AS24818G100 AS24818G25
ACTIAS24818G100EA 0 CHF
ACTIAS24818G100 ACTIAS24818G25
1-Brom-4-propylbenzol
1-Brom-4-propylbenzol
Formel: C₉H₁₁Br
MW: 199,09 g/mol
Siedepunkt: 224 °C (1013 hPa)
Melting Pt: –41.5 °C
Dichte: 1,283 g/cm³ (20 °C)
MDL Number: MFCD00012456
CAS-Nummer: 588-93-2
EINECS: 454-790-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR