4-Ethinylanisol

Lieferant: ACTIVATE SCIENTIFIC

AS32997G1 AS32997G5
ACTIAS32997G1EA 0 CHF
ACTIAS32997G1 ACTIAS32997G5
4-Ethinylanisol
4-Ethinylanisol
Formel: C₉H₈O
MDL Number: MFCD00168815
CAS-Nummer: 768-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR