1-Chlor-4-ethinylbenzol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (4-Chlorophenyl)acetylene, 1-Ethynyl-4-chlorobenzene, (4-Chlorophenyl)ethyne

AS50963G1 AS50963G5
ACTIAS50963G1EA 0 CHF
ACTIAS50963G1 ACTIAS50963G5
1-Chlor-4-ethinylbenzol
1-Chlor-4-ethinylbenzol
Formel: C₈H₅Cl
MW: 136,58 g/mol
MDL Number: MFCD00191917
CAS-Nummer: 873-73-4
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR