2,6-Difluor-4-hydroxybenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS35297G1 AS35297G10 AS35297G5
ACTIAS35297G1EA 0 CHF
ACTIAS35297G1 ACTIAS35297G10 ACTIAS35297G5
2,6-Difluor-4-hydroxybenzoesäure
2,6-Difluoro-4-hydroxybenzoic acid
Formel: C₇H₄F₂O₃
MDL Number: MFCD03094500
CAS-Nummer: 214917-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR