4-Fluor-2-methoxybenzoesäure
Lieferant: ACTIVATE SCIENTIFIC
AS33806G1
AS33806G10
AS33806G5
ACTIAS33806G1EA
0
CHF
ACTIAS33806G1
ACTIAS33806G10
ACTIAS33806G5
4-Fluor-2-methoxybenzoesäure
4-Fluoro-2-methoxybenzoic acid
Formel:
C₈H₇FO₃ |
MDL Number:
MFCD00673010 CAS-Nummer: 395-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...