5-Fluor-2-methoxybenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS35081G1 AS35081G5
ACTIAS35081G1EA 0 CHF
ACTIAS35081G1 ACTIAS35081G5
5-Fluor-2-methoxybenzoesäure
5-Fluor-2-methoxybenzoesäure
Formel: C₈H₇FO₃
MDL Number: MFCD00671765
CAS-Nummer: 394-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR