2-Fluor-6-methoxybenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS33698G1 AS33698G10 AS33698G5
ACTIAS33698G1EA 0 CHF
ACTIAS33698G1 ACTIAS33698G10 ACTIAS33698G5
2-Fluor-6-methoxybenzoesäure
2-Fluoro-6-methoxybenzoic acid
Formel: C₈H₇FO₃
MW: 170,14 g/mol
MDL Number: MFCD00671764
CAS-Nummer: 137654-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR