4-Amino-m-anissäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Amino-3-methoxybenzenecarboxylic acid
AS30955G1
AS30955G5
ACTIAS30955G1EA
0
CHF
ACTIAS30955G1
ACTIAS30955G5
4-Amino-m-anissäure
4-Amino-m-anisic acid
Formel:
C₈H₉NO₃ MW: 167,16 g/mol |
MDL Number:
MFCD00016539 CAS-Nummer: 2486-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...