Ethyl-2-(dimethylamino)benzoat

Lieferant: ACTIVATE SCIENTIFIC

AS36623G1 AS36623G5
ACTIAS36623G1EA 0 CHF
ACTIAS36623G1 ACTIAS36623G5
Ethyl-2-(dimethylamino)benzoat
Ethyl-2-(dimethylamino)benzoat
Formel: C₁₁H₁₅NO₂
MW: 193,242 g/mol
MDL Number: MFCD00009110
CAS-Nummer: 55426-74-9
EINECS: 259-634-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR