4-Cyanbenzolsulfonylchlorid
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Cyanobenzene-1-sulphonyl chloride
AS32869G1
AS32869G5
ACTIAS32869G1EA
0
CHF
ACTIAS32869G1
ACTIAS32869G5
4-Cyanbenzolsulfonylchlorid
4-Cyanbenzolsulfonylchlorid
Formel:
C₇H₄ClNO₂S MW: 201,63 g/mol |
MDL Number:
MFCD00052337 CAS-Nummer: 49584-26-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...