3-Chlorsalicylsäure

Lieferant: ACTIVATE SCIENTIFIC

AS35251G1 AS35251G10 AS35251G5
ACTIAS35251G1EA 0 CHF
ACTIAS35251G1 ACTIAS35251G10 ACTIAS35251G5
3-Chlorsalicylsäure
3-Chlorsalicylsäure
Formel: C₇H₅ClO₃
MW: 172.57 g/mol
Siedepunkt: 320
Melting Pt: 183…183.5 °C
MDL Number: MFCD00010390
CAS-Nummer: 1829-32-9
EINECS: 217-379-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR