3-Chlor-2-methoxybenzoesäure
Lieferant: ACTIVATE SCIENTIFIC
AS33775G1
AS33775G10
AS33775G5
ACTIAS33775G1EA
0
CHF
ACTIAS33775G1
ACTIAS33775G10
ACTIAS33775G5
3-Chlor-2-methoxybenzoesäure
3-Chloro-2-methoxybenzoic acid
Formel:
C₈H₇ClO₃ |
MDL Number:
MFCD00179306 CAS-Nummer: 3260-93-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...