2-Chlor-5-methoxybenzoesäure
Lieferant: ACTIVATE SCIENTIFIC
AS32438G1
AS32438G5
ACTIAS32438G1EA
0
CHF
ACTIAS32438G1
ACTIAS32438G5
2-Chlor-5-methoxybenzoesäure
2-Chlor-5-methoxybenzoesäure
Formel:
C₈H₇ClO₃ |
MDL Number:
MFCD00466261 CAS-Nummer: 6280-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...