1-Brom-3,5-dimethoxybenzol

Lieferant: ACTIVATE SCIENTIFIC

AS24211G100 AS24211G25
ACTIAS24211G100EA 0 CHF
ACTIAS24211G100 ACTIAS24211G25
1-Brom-3,5-dimethoxybenzol
1-Brom-3,5-dimethoxybenzol
Formel: C₈H₉BrO₂
MDL Number: MFCD06200685
CAS-Nummer: 20469-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR