3-Bromphenetol
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Bromophenyl ethyl ether, 1-Bromo-3-ethoxybenzene
AS35321G1
AS35321G5
ACTIAS35321G1EA
0
CHF
ACTIAS35321G1
ACTIAS35321G5
3-Bromphenetol
3-Bromphenetol
Formel:
C₈H₉BrO MW: 201,06 g/mol |
MDL Number:
MFCD00274287 CAS-Nummer: 2655-84-7 EINECS: 220-186-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...