3,4,7,8-Tetramethyl-1,10-phenanthrolin

Lieferant: ACTIVATE SCIENTIFIC

AS21433G1 AS21433G25 AS21433G5
ACTIAS21433G1EA 0 CHF
ACTIAS21433G1 ACTIAS21433G25 ACTIAS21433G5
3,4,7,8-Tetramethyl-1,10-phenanthrolin
3,4,7,8-Tetramethyl-1,10-phenanthroline
Formel: C₁₆H₁₆N₂
MW: 236,32 g/mol
Siedepunkt: 386 °C (1013 hPa)
Melting Pt: 284 °C
MDL Number: MFCD00004974
CAS-Nummer: 1660-93-1
EINECS: 216-762-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR