Scandium(III)chlorid Hexahydrat
Lieferant: ACTIVATE SCIENTIFIC
AS23532G25
AS23532G5
ACTIAS23532G25EA
0
CHF
ACTIAS23532G25
ACTIAS23532G5
Scandium(III)chlorid Hexahydrat
Scandium(III)chlorid Hexahydrat
Formel:
Cl₃Sc·6H₂O |
MDL Number:
MFCD00149852 CAS-Nummer: 20662-14-0 EINECS: 233-799-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...