DL-m-Tyrosin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Hydroxyphenylalanine, 3-(m-Hidroxifenil)-DL-alanina, 3-(3-Hydroxyphenyl)-DL-alanine
AS23049G25
AS23049G5
ACTIAS23049G25EA
86
CHF
ACTIAS23049G25
ACTIAS23049G5
DL-m-Tyrosin
DL-m-Tyrosin
Formel:
C₉H₁₁NO₃ MW: 181,19 g/mol Siedepunkt: 351 °C (1013 hPa) Melting Pt: 283 °C Dichte: 1.456 g/cm³ (20 °C) |
MDL Number:
MFCD00002597 CAS-Nummer: 775-06-4 EINECS: 212-270-9 Merck Index: 12,09971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...