Ytterbium(III)fluorid

Lieferant: ACTIVATE SCIENTIFIC

AS23557G100 AS23557G25
ACTIAS23557G100EA 0 CHF
ACTIAS23557G100 ACTIAS23557G25
Ytterbium(III)fluorid
Ytterbium(III)fluorid
Formel: F₃Yb
Siedepunkt: 2227 °C (1013 hPa)
Melting Pt: 1163,85…1179.85 °C
Dichte: 8,2 g/cm³ (20 °C)
MDL Number: MFCD00049615
CAS-Nummer: 13760-80-0
EINECS: 237-354-2
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR