7-Methylindol

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS50062G100 AS50062G5
ACTIAS50062G100EA 0 CHF
ACTIAS50062G100 ACTIAS50062G5
7-Methylindol
7-Methylindol
Formel: C₉H₉N
Siedepunkt: 266 °C (1013 hPa)
Melting Pt: 83 °C
Dichte: 1,0202 g/cm³ (99.6 °C)
MDL Number: MFCD00005684
CAS-Nummer: 933-67-5
EINECS: 213-270-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR