7-Ethylindol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 7-Ethyl-1H-indole

AS35917G1 AS35917G5
ACTIAS35917G1EA 0 CHF
ACTIAS35917G1 ACTIAS35917G5
7-Ethylindol
7-Ethylindol
Formel: C₁₀H₁₁N
MW: 145,2 g/mol
Siedepunkt: 230 °C (1013 hPa)
Dichte: 1.05 g/cm³ (20 °C)
MDL Number: MFCD00143514
CAS-Nummer: 22867-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR