6-Fluorindol

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 6-Fluoro-1H-indole

AS50080G100 AS50080G5
ACTIAS50080G100EA 0 CHF
ACTIAS50080G100 ACTIAS50080G5
6-Fluorindol
6-Fluorindol
Formel: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 72…77 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00056933
CAS-Nummer: 399-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR