7-Chlorisatin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 7-Chloroindoline-2,3-dione, 7-Chloro-1H-indole-2,3-dione, 7-Cloroindoline-2,3-dione

AS31416G10 AS31416G5
ACTIAS31416G10EA 0 CHF
ACTIAS31416G10 ACTIAS31416G5
7-Chlorisatin
7-Chlorisatin
Formel: C₈H₄ClNO₂
MW: 181,58 g/mol
MDL Number: MFCD00022796
CAS-Nummer: 7477-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR