Indol-5-carbonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1H-Indole-5-carbonitrile, 5-Cyanoindole, 5-Cyano-1H-indole

AS7031G100 AS7031G5
ACTIAS7031G100EA 0 CHF
ACTIAS7031G100 ACTIAS7031G5
Indol-5-carbonitril
Indol-5-carbonitril
Formel: C₉H₆N₂
MW: 142.16 g/mol
Melting Pt: 106…108 °C
MDL Number: MFCD00005669
CAS-Nummer: 15861-24-2
EINECS: 239-986-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR