6-Fluorisatin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
6-Fluoro-1H-indole-2,3-dione, 6-Fluoroindoline-2,3-dione
AS35909G1
AS35909G5
ACTIAS35909G1EA
0
CHF
ACTIAS35909G1
ACTIAS35909G5
6-Fluorisatin
6-Fluorisatin
Formel:
C₈H₄FNO₂ MW: 165,12 g/mol |
MDL Number:
MFCD03618556 CAS-Nummer: 324-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...