4-Nitrophenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
AS50896G1
AS50896G5
ACTIAS50896G1EA
0
CHF
ACTIAS50896G1
ACTIAS50896G5
4-Nitrophenylboronsäure
4-Nitrophenylboronsäure
Formel:
C₆H₆BNO₄ |
MDL Number:
MFCD00161360 CAS-Nummer: 24067-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...