4-Methoxyphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
4-Methoxybenzeneboronic acid, Dihydroxy(4-methoxyphenyl)borane, (4-Methoxyphenyl)metaboric acid
AS20118G100
AS20118G25
ACTIAS20118G100EA
68
CHF
ACTIAS20118G100
ACTIAS20118G25
4-Methoxyphenylboronsäure
4-Methoxyphenylboronsäure
Formel:
C₇H₉BO₃ MW: 151,96 g/mol Melting Pt: 204…207 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00039139 CAS-Nummer: 5720-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...