4-Chlor-2-hydroxypyridin
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
AS31281G1
AS31281G5
ACTIAS31281G1EA
0
CHF
ACTIAS31281G1
ACTIAS31281G5
4-Chlor-2-hydroxypyridin
4-Chlor-2-hydroxypyridin
Formel:
C₅H₄ClNO MW: 129,55 g/mol |
MDL Number:
MFCD03093022 CAS-Nummer: 40673-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...