4-Brom-2-methoxypyridin
Lieferant: ACTIVATE SCIENTIFIC
AS50476G1
AS50476G5
ACTIAS50476G1EA
0
CHF
ACTIAS50476G1
ACTIAS50476G5
4-Brom-2-methoxypyridin
4-Bromo-2-methoxypyridine
Formel:
C₆H₆BrNO |
MDL Number:
MFCD08277268 CAS-Nummer: 100367-39-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...