4-Brom-2-fluorpyridin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS1008G1 AS1008G5
ACTIAS1008G1EA 0 CHF
ACTIAS1008G1 ACTIAS1008G5
4-Brom-2-fluorpyridin
4-Bromo-2-fluoropyridine
Formel: C₅H₃BrFN
MW: 175,99 g/mol
MDL Number: MFCD04112504
CAS-Nummer: 128071-98-7
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR