4-Brom-2-chlorpyridin

Lieferant: ACTIVATE SCIENTIFIC

AS50349G100 AS50349G5
ACTIAS50349G100EA 0 CHF
ACTIAS50349G100 ACTIAS50349G5
4-Brom-2-chlorpyridin
4-Bromo-2-chloropyridine
Formel: C₅H₃BrClN
MW: 192,44 g/mol
MDL Number: MFCD03840756
CAS-Nummer: 73583-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR