4-Amino-3-nitropyridin
Lieferant: ACTIVATE SCIENTIFIC
|
Achtung
|
Synonyms:
3-Nitropyridin-4-amine, 3-Nitro-4-pyridylamine, 3-Nitropyridin-4-ylamine
AS22744G25
AS22744G5
ACTIAS22744G25EA
65
CHF
ACTIAS22744G25
ACTIAS22744G5
4-Amino-3-nitropyridin
4-Amino-3-nitropyridin
Formel:
C₅H₅N₃O₂ MW: 139,11 g/mol |
MDL Number:
MFCD00160304 CAS-Nummer: 1681-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...