4-Amino-3-methylpyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Amino-3-picoline, 3-Methyl-4-pyridylamine, 3-Methylpyridin-4-amine

AS7071G100 AS7071G5
ACTIAS7071G100EA 0 CHF
ACTIAS7071G100 ACTIAS7071G5
4-Amino-3-methylpyridin
4-Amino-3-methylpyridin
Formel: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD01704431
CAS-Nummer: 1990-90-5
EINECS: 217-872-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR