4-Amino-2,6-dichlorpyridin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 2,6-Dichloro-4-pyridylamine, 2,6-Dichloropyridin-4-amine

AS31382G1 AS31382G5
ACTIAS31382G1EA 0 CHF
ACTIAS31382G1 ACTIAS31382G5
4-Amino-2,6-dichlorpyridin
4-Amino-2,6-dichloropyridine
Formel: C₅H₄Cl₂N₂
MW: 163,01 g/mol
MDL Number: MFCD00052832
CAS-Nummer: 2587-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR