4-(Difluormethoxy)benzaldehyd
Lieferant: ACTIVATE SCIENTIFIC
AS34363G1
AS34363G10
AS34363G5
ACTIAS34363G1EA
0
CHF
ACTIAS34363G1
ACTIAS34363G10
ACTIAS34363G5
4-(Difluormethoxy)benzaldehyd
4-(Difluormethoxy)benzaldehyd
Formel:
C₈H₆F₂O₂ MW: 172,13 g/mol |
MDL Number:
MFCD00042252 CAS-Nummer: 73960-07-3 EINECS: 277-653-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...