3-Fluor-5-nitrobenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS33702G1 AS33702G5
ACTIAS33702G1EA 0 CHF
ACTIAS33702G1 ACTIAS33702G5
3-Fluor-5-nitrobenzoesäure
3-Fluor-5-nitrobenzoesäure
Formel: C₇H₄FNO₄
MDL Number: MFCD05864360
CAS-Nummer: 14027-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR