3-Fluor-5-hydroxypyridin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Fluoro-5-pyridinol, 5-Fluoropyridin-3-ol

AS30507G1 AS30507G5
ACTIAS30507G1EA 0 CHF
ACTIAS30507G1 ACTIAS30507G5
3-Fluor-5-hydroxypyridin
3-Fluor-5-hydroxypyridin
Formel: C₅H₄FNO
MW: 113,09 g/mol
MDL Number: MFCD03095199
CAS-Nummer: 209328-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR