3-Fluor-4-iodpyridin
Lieferant: ACTIVATE SCIENTIFIC
|
Gefahr
|
AS35810G1
AS35810G5
ACTIAS35810G1EA
0
CHF
ACTIAS35810G1
ACTIAS35810G5
3-Fluor-4-iodpyridin
3-Fluor-4-iodpyridin
Formel:
C₅H₃FIN Lager Temperatur: Kühlschrank |
MDL Number:
MFCD03092927 CAS-Nummer: 22282-75-3 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...