3-Chlorphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC GMBH
Achtung

Synonyms: (3-Chlorphenyl)metaborsäure3-ChlorbenzolboronsäureDihydroxy(3-chlorphenyl)boran

ACTIAS50130G5EA 0 CHF
ACTIAS50130G5 ACTIAS50130G1 ACTIAS50130G25
3-Chlorphenylboronsäure
3-Chlorphenylboronsäure
Formel: C₆H₆BClO₂
MW: 156,38 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00161354
CAS-Nummer: 63503-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR