5-Chlor-2-fluorbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC
Achtung

AS31945G10 AS31945G5
ACTIAS31945G10EA 0 CHF
ACTIAS31945G10 ACTIAS31945G5
5-Chlor-2-fluorbenzoesäure
5-Chlor-2-fluorbenzoesäure
Formel: C₇H₄ClFO₂
MDL Number: MFCD00665762
CAS-Nummer: 394-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR