5-Chlor-3-hydroxypyridin

Lieferant: ACTIVATE SCIENTIFIC GMBH
Achtung

Synonyms: 3-Chlor-5-pyridinol5-Chlor-3-pyridinol3-Chlor-5-hydroxypyridin

ACTIAS31602G1EA 0 CHF
ACTIAS31602G1 ACTIAS31602G10 ACTIAS31602G5
5-Chlor-3-hydroxypyridin
5-Chlor-3-hydroxypyridin
Formel: C₅H₄ClNO
MW: 129,55 g/mol
Melting Pt: 158 °C
MDL Number: MFCD00006379
CAS-Nummer: 74115-12-1
EINECS: 277-727-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR