3-Chlor-2-hydroxypyridin

Lieferant: ACTIVATE SCIENTIFIC
Achtung

Synonyms: 3-Chloro-2-pyridinol

AS22691G25 AS22691G5
ACTIAS22691G25EA 0 CHF
ACTIAS22691G25 ACTIAS22691G5
3-Chlor-2-hydroxypyridin
3-Chlor-2-hydroxypyridin
Formel: C₅H₄ClNO
MW: 129,55 g/mol
MDL Number: MFCD01646109
CAS-Nummer: 13466-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR