5-Brom-2-methylpyridin
Lieferant: ACTIVATE SCIENTIFIC
AS20196G100
AS20196G25
ACTIAS20196G100EA
0
CHF
ACTIAS20196G100
ACTIAS20196G25
5-Brom-2-methylpyridin
5-Brom-2-methylpyridin
Formel:
C₆H₆BrN |
MDL Number:
MFCD00661170 CAS-Nummer: 3430-13-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...